Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102448.4 | NP_174006.2 | 5 - 6 | 615 | 76 | |
GenBank mRNA | AJ575584.1 | - | 5 - 6 | 463 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |