Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCT1;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332773.1 | NP_001321314.1 | 6 - 7 | 1581 | 86 | |
NM_001332774.1 | NP_001321315.1 | 5 - 6 | 1691 | 86 | ||
NM_001332776.1 | NP_001321317.1 | 6 - 7 | 1360 | 86 | ||
NM_001332777.1 | NP_001321318.1 | 4 - 5 | 1255 | 86 | ||
NM_102528.4 | NP_174084.1 | 6 - 7 | 1028 | 86 | ||
GenBank mRNA | AF344323.1 | - | 5 - 6 | 772 | 86 | |
AK117692.1 | - | 6 - 7 | 978 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |