Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344021.1 | NP_001331944.1 | 10 - 11 | 1271 | 65 | |
NM_001344022.1 | NP_001331946.1 | 10 - 11 | 1271 | 65 | ||
NM_001344023.1 | NP_001331945.1 | 10 - 11 | 1102 | 65 | ||
NM_122645.5 | NP_198115.2 | 10 - 11 | 1128 | 65 | ||
GenBank mRNA | AY093156.1 | - | 10 - 11 | 1102 | 65 | |
BT008399.1 | - | 10 - 11 | 1024 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |