Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123358.4 | NP_198811.1 | 1 - 2 | 242 | 71 | |
GenBank mRNA | AY048235.1 | - | 1 - 2 | 156 | 71 | |
AY113052.1 | - | 1 - 2 | 145 | 71 | ||
Z35475.1 | - | 163 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |