Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123384.3 | NP_568577.1 | 7 - 8 | 1823 | 126 | |
GenBank mRNA | AY046023.1 | - | 7 - 8 | 1639 | 126 | |
AY142608.1 | - | 7 - 8 | 1567 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |