Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123506.3 | NP_198957.1 | 2 - 3 | 1733 | 86 | |
GenBank mRNA | AY049237.1 | - | 2 - 3 | 1542 | 86 | |
BT002637.1 | - | 2 - 3 | 1245 | 86 | ||
U39944.2 | - | 2 - 3 | 1625 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Arabidopsis thaliana Probe spans exons |