Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344537.1 | NP_001331906.1 | 2 - 3 | 754 | 74 | |
NM_123786.4 | NP_199232.1 | 2 - 3 | 735 | 74 | ||
GenBank mRNA | AB062490.1 | - | 2 - 3 | 581 | 74 | |
AY026773.1 | - | 2 - 3 | 581 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |