Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123990.3 | NP_568658.1 | 13 - 14 | 2183 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |