Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344713.1 | NP_001331621.1 | 2 - 3 | 685 | 121 | |
NM_124095.4 | NP_568680.1 | 2 - 3 | 637 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |