Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124481.4 | NP_199915.1 | 3 - 4 | 434 | 102 | |
GenBank mRNA | AB120415.1 | - | 3 - 4 | 371 | 102 | |
AF370196.1 | - | 3 - 4 | 374 | 102 | ||
AY113932.1 | - | 3 - 4 | 294 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |