Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124635.4 | NP_568775.1 | 8 - 9 | 1069 | 88 | |
GenBank mRNA | AF360236.1 | - | 8 - 9 | 932 | 88 | |
AY040035.1 | - | 7 - 8 | 746 | 88 | ||
DQ324529.1 | - | 7 - 8 | 746 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |