Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202755.2 | NP_001189684.1 | 7 - 8 | 888 | 81 | |
NM_001336565.1 | NP_001324113.1 | 7 - 8 | 1081 | 81 | ||
NM_001336566.1 | NP_001324112.1 | 7 - 8 | 827 | 81 | ||
NM_129116.5 | NP_181105.2 | 7 - 8 | 852 | 81 | ||
GenBank mRNA | FJ708709.1 | - | 6 - 7 | 527 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |