Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084585.2 | NP_001078054.1 | 4 - 5 | 456 | 143 | |
NM_129973.3 | NP_181938.1 | 4 - 5 | 679 | 143 | ||
GenBank mRNA | AF361801.1 | - | 4 - 5 | 446 | 143 | |
AK226690.1 | - | 4 - 5 | 449 | 143 | ||
AK227643.1 | - | 4 - 5 | 446 | 143 | ||
AK316933.1 | - | 4 - 5 | 450 | 143 | ||
AY050418.2 | - | 4 - 5 | 447 | 143 | ||
AY113036.1 | - | 4 - 5 | 344 | 143 | ||
D83531.1 | - | 4 - 5 | 447 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |