Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201908.1 | NP_973637.1 | 8 - 9 | 1210 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |