Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2A-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115712.4 | NP_567066.1 | 3 - 4 | 464 | 137 | |
GenBank mRNA | AY056222.1 | - | 3 - 4 | 430 | 137 | |
AY057604.1 | - | 3 - 4 | 414 | 137 | ||
AY113023.1 | - | 3 - 4 | 239 | 137 | ||
U08047.1 | - | 3 - 4 | 358 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |