Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ProT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336602.1 | NP_001324136.1 | 5 - 6 | 1220 | 150 | |
NM_129215.4 | NP_181198.1 | 5 - 6 | 1220 | 150 | ||
GenBank mRNA | BT004018.1 | - | 5 - 6 | 809 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |