Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123832.2 | NP_001117304.1 | 5 | 1580 | 74 | |
NM_101691.9 | NP_173269.1 | 6 | 1698 | 74 | ||
GenBank mRNA | AY063952.1 | - | 6 | 1357 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |