Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115390.3 | NP_191092.1 | 11 | 4347 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |