Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103312.1 | NP_001096782.1 | 1 - 2 | 209 | 77 | |
GenBank mRNA | BC149939.1 | - | 1 - 2 | 209 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |