Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034799.1 | NP_001029971.1 | 1 - 2 | 495 | 105 | |
GenBank mRNA | BC103144.1 | - | 1 - 2 | 495 | 105 | |
BT026259.1 | - | 1 - 2 | 375 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |