Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040471.1 | NP_001035561.1 | 8 - 9 | 834 | 94 | |
GenBank mRNA | AF043228.1 | - | 1 - 2 | 60 | 94 | |
BC103416.1 | - | 8 - 9 | 834 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |