Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174264.3 | NP_776689.2 | 6 - 7 | 979 | 71 | |
GenBank mRNA | AF080219.1 | - | 6 - 7 | 879 | 71 | |
BC118382.1 | - | 6 - 7 | 886 | 71 | ||
BT020648.1 | - | 6 - 7 | 979 | 71 | ||
BT020669.1 | - | 6 - 7 | 951 | 71 | ||
BT020822.1 | - | 6 - 7 | 931 | 71 | ||
BT021153.1 | - | 6 - 7 | 939 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |