Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174288.1 | NP_776713.1 | 3 - 4 | 586 | 154 | |
GenBank mRNA | U02292.1 | - | 3 - 4 | 586 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |