Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205777.2 | NP_991346.2 | 3 - 4 | 293 | 76 | |
NM_205818.2 | NP_991387.2 | 3 - 4 | 340 | 76 | ||
GenBank mRNA | AY911378.1 | - | 3 - 4 | 76 | ||
BC126769.1 | - | 3 - 4 | 272 | 76 | ||
BC148015.1 | - | 3 - 4 | 307 | 76 | ||
CB454302.1 | - | 2 - 3 | 441 | 76 | ||
CB455704.1 | - | 3 - 4 | 302 | 76 | ||
CB463455.1 | - | 2 - 3 | 240 | 76 | ||
CR454063.1 | - | 3 - 4 | 321 | 76 | ||
X92956.1 | - | 279 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |