Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205817.1 | NP_991386.1 | 10 - 11 | 978 | 90 | |
GenBank mRNA | BC102359.1 | - | 10 - 11 | 980 | 90 | |
BT025401.1 | - | 10 - 11 | 932 | 90 | ||
X59418.1 | - | 978 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |