Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015559.1 | NP_001015559.1 | 8 - 9 | 1059 | 61 | |
GenBank mRNA | BT020833.1 | - | 8 - 9 | 1059 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |