Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015577.1 | NP_001015577.1 | 3 - 4 | 439 | 133 | |
GenBank mRNA | BT020881.1 | - | 3 - 4 | 439 | 133 | |
BT021707.1 | - | 3 - 4 | 430 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |