Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC51A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025333.2 | NP_001020504.1 | 4 - 5 | 507 | 75 | |
GenBank mRNA | AY911312.1 | - | 4 - 5 | 508 | 75 | |
BC102154.1 | - | 4 - 5 | 551 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |