Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030304.1 | NP_001025475.1 | 3 - 4 | 870 | 60 | |
GenBank mRNA | AM048630.1 | - | 3 - 4 | 870 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |