Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRXL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040598.1 | NP_001035688.1 | 1 - 2 | 147 | 100 | |
GenBank mRNA | BC114900.1 | - | 1 - 2 | 66 | 100 | |
BT021811.1 | - | 1 - 2 | 147 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |