Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174121.1 | NP_776546.1 | 1 - 2 | 1136 | 74 | |
GenBank mRNA | BC118240.1 | - | 1 - 2 | 1130 | 74 | |
U83919.1 | - | 1 - 2 | 1153 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |