Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174654.2 | NP_777079.1 | 13 - 14 | 1947 | 87 | |
GenBank mRNA | AJ508718.1 | - | 13 - 14 | 1947 | 87 | |
AY052775.1 | - | 14 - 15 | 2031 | 87 | ||
BC133437.1 | - | 13 - 14 | 1992 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |