Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176633.3 | NP_788806.1 | 2 - 3 | 1014 | 126 | |
GenBank mRNA | AB003485.1 | - | 2 - 3 | 1016 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |