Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034372.1 | NP_001029544.1 | 2 - 3 | 252 | 74 | |
GenBank mRNA | BC103078.1 | - | 2 - 3 | 252 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |