Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034728.1 | NP_001029900.1 | 6 - 7 | 952 | 80 | |
GenBank mRNA | BC104630.1 | - | 6 - 7 | 952 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |