Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035349.1 | NP_001030426.1 | 6 - 7 | 622 | 112 | |
GenBank mRNA | BC104604.1 | - | 6 - 7 | 622 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |