Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANGRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037460.2 | NP_001032537.1 | 4 - 5 | 559 | 66 | |
GenBank mRNA | BC108151.1 | - | 4 - 5 | 560 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |