Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037476.2 | NP_001032553.1 | 3 - 4 | 1627 | 66 | |
GenBank mRNA | BC108094.1 | - | 3 - 4 | 1628 | 66 | |
BT026530.1 | - | 3 - 4 | 1635 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |