Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037488.2 | NP_001032565.1 | 4 - 5 | 569 | 108 | |
GenBank mRNA | BC108118.1 | - | 4 - 5 | 570 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |