Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C13H20orf85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075198.2 | NP_001068666.1 | 3 - 4 | 310 | 73 | |
GenBank mRNA | BC109834.1 | - | 3 - 4 | 312 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |