Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076467.1 | NP_001069935.1 | 4 - 5 | 412 | 84 | |
NM_001370742.1 | NP_001357671.1 | 4 - 5 | 370 | 84 | ||
GenBank mRNA | BC102743.1 | - | 4 - 5 | 412 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |