Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077046.1 | NP_001070514.1 | 4 - 5 | 431 | 94 | |
GenBank mRNA | BC104531.1 | - | 4 - 5 | 431 | 94 | |
BT025413.1 | - | 4 - 5 | 409 | 94 | ||
BT026183.1 | - | 4 - 5 | 413 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |