Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024509.3 | NP_001019680.2 | 10 - 11 | 781 | 69 | |
GenBank mRNA | BC105379.1 | - | 10 - 11 | 782 | 69 | |
DQ010410.1 | - | 10 - 11 | 699 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |