Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038090.2 | NP_001033179.1 | 5 - 6 | 817 | 74 | |
GenBank mRNA | BC109917.1 | - | 5 - 6 | 818 | 74 | |
BC148921.1 | - | 4 - 5 | 1366 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |