Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038188.2 | NP_001033277.1 | 3 - 4 | 1668 | 101 | |
GenBank mRNA | BC109654.1 | - | 3 - 4 | 1669 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |