Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038512.2 | NP_001033601.1 | 4 - 5 | 1515 | 113 | |
GenBank mRNA | BC109844.1 | - | 4 - 5 | 1516 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |