Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038542.2 | NP_001033631.1 | 4 - 5 | 457 | 70 | |
GenBank mRNA | BC109846.1 | - | 4 - 5 | 458 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |