Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046140.2 | NP_001039605.1 | 4 - 5 | 813 | 143 | |
GenBank mRNA | BC105395.1 | - | 4 - 5 | 814 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |