Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079619.2 | NP_001073087.1 | 3 - 4 | 270 | 92 | |
GenBank mRNA | BC112592.1 | - | 3 - 4 | 271 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |