Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034632.3 | NP_001029804.1 | 2 - 3 | 285 | 96 | |
GenBank mRNA | BC112583.1 | - | 2 - 3 | 287 | 96 | |
BT021189.1 | - | 2 - 3 | 276 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |